Anti-ZBTB7 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-10235R-CY5
BOSSBS-10235R-CY5EA 900 EUR
BOSSBS-10235R-CY5
Anti-ZBTB7 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Pokemon, the POK erythroid myeloid ontogenic factor, not only regulates the expression of many genes, but also plays an important role in cell tumorigenesis. To investigate the molecular mechanism regulating expression of the Pokemon gene in humans, its 5'-upstream region was cloned and analyzed. Transient analysis revealed that the Pokemon promoter is constitutive. Deletion analysis and a DNA decoy assay indicated that the NEG-U and NEG-D elements were involved in negative regulation of the Pokemon promoter, whereas the POS-D element was mainly responsible for its strong activity.More Product Information