Anti-C2a Rabbit Polyclonal Antibody
Supplier: Bioss
BS-10428R
BOSSBS-10428REA 704 EUR
BOSSBS-10428R
Anti-C2a Rabbit Polyclonal Antibody
Antibodies
Component C2 is a serum glycoprotein that functions as part of the classical pathway of the complement system. Activated C1 cleaves C2 into C2a and C2b. The serine proteinase C2a then combines with complement factor 4b to create the C3 or C5 convertase. Deficiency of C2 has been reported to associated with certain autoimmune diseases and SNPs in this gene have been associated with altered susceptibility to age-related macular degeneration. This gene localizes within the class III region of the MHC on the short arm of chromosome 6. Alternative splicing results in multiple transcript variantsMore Product Information