Anti-JAM3 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-11086R-CY3
BOSSBS-11086R-CY3EA 900 EUR
BOSSBS-11086R-CY3
Anti-JAM3 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Participates in cell-cell adhesion. It is a counter-receptor for ITGAM, mediating leukocyte-platelet interactions and is involved in the regulation of transepithelial migration of polymorphonuclear neutrophils (PMN). The soluble form is a mediator of angiogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JAM3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR