Anti-SV2B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-11365R-A555
BOSSBS-11365R-A555EA 893 EUR
BOSSBS-11365R-A555
Anti-SV2B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
In all vertebrates, SV2 proteins are abundant, hydrophobic, membrane glycoproteins that are expressed as two major isoforms, SV2A and SV2B, and one minor isoform, SV2C. SV2 proteins are differentially expressed in the brain and are present on all synaptic vesicles, independent of transmitter type. SV2A is abundantly expressed in the subcortex, specifically in the synaptic vesicles of all presynaptic nerve terminals, and also in most neuroendocrine secretory granules. SV2B displays a more restricted pattern of expression in that it is only present on a small subset of synapses in theMore Product Information