Anti-PTPRF Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-11530R-CY3
BOSSBS-11530R-CY3EA 900 EUR
BOSSBS-11530R-CY3
Anti-PTPRF Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The first PTPase domain has enzymatic activity, while the second one seems to affect the substrate specificity of the first one.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPRF
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR