Anti-SOX8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-11600R-A555
BOSSBS-11600R-A555EA 893 EUR
BOSSBS-11600R-A555
Anti-SOX8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of thisMore Product Information