Anti-NAV3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-12151R-A350
BOSSBS-12151R-A350EA 893 EUR
BOSSBS-12151R-A350
Anti-NAV3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
May regulate IL2 production by T-cells. May be involved in neuron regeneration.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NAV3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol NAV3
Environmentally Preferable
Antigen name Neuron Navigator 3
Antigen synonyms KIAA0938|POMFIL1|STEERIN3|Neuron navigator 3|Pore membrane and/or filament-interacting-like protein 1|unc53H3|Steerin-3|Unc-53 homolog 3
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR