Anti-ABCB6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-1224R-A555
BOSSBS-1224R-A555EA 893 EUR
BOSSBS-1224R-A555
Anti-ABCB6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate geneMore Product Information