Anti-VNN2 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-12340R
BOSSBS-12340REA 725 EUR
BOSSBS-12340R
Anti-VNN2 Rabbit Polyclonal Antibody
Antibodies
Vanin-2 is a 520 amino acid GPI-anchor protein that belongs to the CN hydrolase family and BTD/VNN subfamily.Widely expressed with high expression in spleen and blood, vanin-2 is suggested to be involved in thymus homing of bone marrow cells, regulation of Integrin ∫2-mediated cell adhesion, and migration and motility of neutrophils. Vanin-2 exists as five alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 6q23.2.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: VNN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR