Anti-ICAM2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-1258R-CY7
BOSSBS-1258R-CY7EA 900 EUR
BOSSBS-1258R-CY7
Anti-ICAM2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). ICAM2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ICAM2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR