Anti-CRYBA1 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-12859R-CY3
BOSSBS-12859R-CY3EA 900 EUR
BOSSBS-12859R-CY3
Anti-CRYBA1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Crystallins are the major proteins of the vertebrate eye lens, where they maintain the transparency and refractive index of the lens. Crystallins are divided into a, b, and g families, and the b- and g-crystallins also comprise a superfamily. Crystallins usually contain seven distinctive protein regions, including four homologous motifs, a connecting peptide, and N- and C-terminal extensions. b-crystallins constitute the major lens structural proteins. They associate into dimers, tetramers, and higher order aggregates. The b-crystallin subfamily is composed of several gene products,More Product Information