Anti-NFKBIA Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-1287R-CY7
BOSSBS-1287R-CY7EA 909 EUR
BOSSBS-1287R-CY7
Anti-NFKBIA Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localization signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NFKBIA/IKB alpha
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR