Anti-CHRM3 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-1289R
BOSSBS-1289REA 752 EUR
BOSSBS-1289R
Anti-CHRM3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CHRM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR