Anti-DKC1 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-13007R-CY7
BOSSBS-13007R-CY7EA 900 EUR
BOSSBS-13007R-CY7
Anti-DKC1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Dyskerin (NAP57) associates with the chaperone protein Nopp140 and forms a small ribonucleoprotein particle with GAR1 (NOLA1), NHP2 (NOLA2) and Nop10 for the isomerization of uridine to pseudouridine (1). GAR1, NHP2 and dyskerin localize to the dense fibrillar component of the nucleolus and in nuclear Cajal bodies (2). The dyskerin gene maps to chromosome Xq28 (3). Missense mutations in the dyskerin gene interfere with normal nuclear localization of dyskerin and cause Dyskeratosis congenita (DKC) (4). DKC is a rare, X-linked bone marrow disorder characterized by cutaneous hyperpigmentation,More Product Information