Anti-Complement component C9b Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Supplier: Bioss
BS-15307R-A647
BOSSBS-15307R-A647EA 893 EUR
BOSSBS-15307R-A647
Anti-Complement component C9b Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Antibodies
C9 is a plasma protein synthesised in the liver and monocytes consisting of a single polypeptide chain. C9 is a part of the membrane attack complex (MAC), an important component of the immune system. The MAC forms upon complement system activation by invading pathogenic bacteria and consists of the four major complement proteins: C5b, C6, C7 and C8. These complement proteins bind to the outer surface of the plasma membrane of the invading cell. C9 binds to the membrane associated C5b-8 protein, which leads to the circular polymerisation of 12-18 C9 molecules. These polymerised C9 moleculesMore Product Information