Anti-GPCR C5L2 Rabbit Polyclonal Antibody (HRP)

Supplier: Bioss
BS-15352R-HRP
BOSSBS-15352R-HRPEA 900 EUR
BOSSBS-15352R-HRP
Anti-GPCR C5L2 Rabbit Polyclonal Antibody (HRP)
Antibodies
GPCR C5L2.

Type: Primary
Antigen: GPCR C5L2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 1-100/337
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR