Anti-Hamartin Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Supplier: Bioss
BS-15406R-A680
BOSSBS-15406R-A680EA 890 EUR
BOSSBS-15406R-A680
Anti-Hamartin Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antibodies
Tuberous sclerosis complex (TSC) is an autosomal dominant genetic disorder characterised by mental retardation and the widespread development of distinctive tumors termed hamartomas. Two different genetic loci have been linked to TSC; one of these loci, the tuberous sclerosis-2 gene (TSC2), encodes a protein called tuberin and the other loci, tuberous sclerosis-1 gene (TSC1), encodes a protein called hamartin. Tuberin and hamartin interact with each other forming a cystoplasmic complex. Hamartin interacts with the ezrin-radixin-moesin (ERM) family of actin-binding proteins and inhibition ofMore Product Information