Anti-T140 Rabbit Polyclonal Antibody
Supplier: Bioss
BS-15559R
BOSSBS-15559REA 725 EUR
BOSSBS-15559R
Anti-T140 Rabbit Polyclonal Antibody
Antibodies
IFT140 is a gene encodes one of the subunits of the intraflagellar transport (IFT) complex A. Intraflagellar transport is involved in the genesis, resorption and signaling of primary cilia. The primary cilium is a microtubule-based sensory organelle at the surface of most quiescent mammalian cells, that receives signals from its environment, such as the flow of fluid, light or odors, and transduces those signals to the nucleus. Loss of the corresponding protein in mouse results in renal cystic disease.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: T140