Anti-ERK1 Thr202/Tyr204+ERK2 Thr183/Tyr185 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-1646R-A680
BOSSBS-1646R-A680EA 917 EUR
BOSSBS-1646R-A680
Anti-ERK1 Thr202/Tyr204+ERK2 Thr183/Tyr185 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets. Two alternatively spliced transcript variantsMore Product Information