Anti-STAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-1657R-A488
BOSSBS-1657R-A488EA 898 EUR
BOSSBS-1657R-A488
Anti-STAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein can be activated by various ligands including interferon-alpha, interferon-gamma, EGF, PDGF and IL6. This protein mediates the expression of a variety of genes, which is thought to be important for cell viability in response to different cell stimuli and pathogens. TwoMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol STAT1
Environmentally Preferable
Antigen name Signal Transducer And Activator of Transcription 1
Antigen synonyms signal transducer and activator of transcription 1|STAT91|ISGF-3|IMD31B|CANDF7|IMD31C|IMD31A
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 6772
Isotype IgG
ImmunoChemistry Yes
Modification phospho-Tyr701
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR