Anti-p-CK Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Supplier: Bioss
BS-1712R-A488
BOSSBS-1712R-A488EA 901 EUR
BOSSBS-1712R-A488
Anti-p-CK Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Cytokeratins are proteins of keratin-containing intermediate filaments found in the intracytoplasmic cytoskeleton of epithelial tissue. The cytokeratins are encoded by a family encompassing 30 genes. Among them, 20 are epithelial genes and the remaining 10 are specific for trichocytes.
In the cytoplasm, the keratin filaments conform a complex network which extends from the surface of the nucleus to the cell membrane. Numerous accessory proteins are involved in the genesis and maintenance of such structure. This association between the plasma membrane and the nuclear surface provides importantMore Product Information