Anti-GPAT4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-1924R-A488
BOSSBS-1924R-A488EA 893 EUR
BOSSBS-1924R-A488
Anti-GPAT4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
AGPAT6 is a novel glycerolipid acyltransferase of the ER, which is crucial for the production of milk fat by the mammary gland. It converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSARG7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR