Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2197R
BOSSBS-2197REA 725 EUR
BOSSBS-2197R
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: duck hepatitis A virus 1 Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR