Anti-CSF1R Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-2755R-CY5.5
BOSSBS-2755R-CY5.5EA 909 EUR
BOSSBS-2755R-CY5.5
Anti-CSF1R Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The protein encoded by this gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most if not all of the biological effects of this cytokine. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in this gene have been associated with a predisposition to myeloid malignancy. The firstMore Product Information