Anti-VIM Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-3472R-CY5.5
BOSSBS-3472R-CY5.5EA 898 EUR
BOSSBS-3472R-CY5.5
Anti-VIM Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cellMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol VIM
Antigen name Vimentin
Environmentally Preferable
Antigen synonyms CTRCT30|HEL113
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 7431
Isotype IgG
ImmunoChemistry Yes
Modification phospho-Ser82
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR