Anti-MTF1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-3601R-CY5
BOSSBS-3601R-CY5EA 900 EUR
BOSSBS-3601R-CY5
Anti-MTF1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Activates the metallothionein I promoter. Binds to the metal responsive element (MRE).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR