Anti-SCD Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-3787R-A488
BOSSBS-3787R-A488EA 893 EUR
BOSSBS-3787R-A488
Anti-SCD Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Terminal component of the liver microsomal stearyl-CoA desaturase system, that utilizes O(2) and electrons from reduced cytochrome b5 to catalyze the insertion of a double bond into a spectrum of fatty acyl-CoA substrates including palmitoyl-CoA and stearoyl-CoA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR