Anti-AMY2A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-4030R-FITC
BOSSBS-4030R-FITCEA 900 EUR
BOSSBS-4030R-FITC
Anti-AMY2A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Pancreatic amylase is a digestive enzyme secreted by the pancreas. It consists of a single polypeptide chain with a molecular weight of 54 kDa.Pancreatic amylase and salivary amylase are the products of separate genes and, although they share 98% amino acid sequence homology, they differ in molecular size, isoelectric point and antigenic properties. Measurement of amylase concentrations in serum is a diagnostic test for various pancreatic disorders (especially acute and relapsing pancreatitis).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMY2A
Clonality: Polyclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR