Anti-SLC25A12 Rabbit Polyclonal Antibody
Supplier: Bioss
BS-4038R
BOSSBS-4038REA 725 EUR
BOSSBS-4038R
Anti-SLC25A12 Rabbit Polyclonal Antibody
Antibodies
SLC25A13 is a member of the mitochondrial carrier family. It contains four EF-hand Ca(2+) binding motifs in the N-terminal domain, and localizes to mitochondria. It catalyzes the exchange of aspartate for glutamate and a proton across the inner mitochondrial membrane, and is stimulated by calcium on the external side of the inner mitochondrial membrane. Mutations in the SLC25A13 gene result in citrullinemia, type II. Multiple transcript variants encoding different isoforms have been found for this gene.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: SLC25A12More Product Information