Anti-TBL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-4219R-A680
BOSSBS-4219R-A680EA 890 EUR
BOSSBS-4219R-A680
Anti-TBL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. This encoded protein is found as a subunit in corepressor SMRT (silencing mediator for retinoid and thyroidMore Product Information