Anti-DEFB4A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-4307R-HRP
BOSSBS-4307R-HRPEA 883 EUR
BOSSBS-4307R-HRP
Anti-DEFB4A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Beta-defensins (also designated BD, and hBD in human) are small cationic peptides with broad-spectrum antimicrobial activity. Produced in mucosal epithelia and neutrophils of several species, Beta-defensins are developmentally regulated. Human b-defensin 2 is locally regulated by inflammation and is the first member of the b-defensin family that is locally inducible by inflammation. The murine homolog of human b-defensin 2, which is called b-defensin 3, is present in the respiratory system and in low levels in the epithelial cells of the intestine and lung. The unique murine b-defensin 2 (More Product Information
Order Now

SPECIFICATIONS

Antigen symbol DEFB4A
Environmentally Preferable
Antigen name Defensin, beta 4A
Antigen synonyms DEFB2|BD-2|SAP1|DEFB4|DEFB-2|DEFB102|HBD-2
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Gene ID 1673
Isotype IgG
ImmunoChemistry Yes
Immunogen 34-64/64
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR