Anti-CTAG1B Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-4501R-CY7
BOSSBS-4501R-CY7EA 900 EUR
BOSSBS-4501R-CY7
Anti-CTAG1B Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The protein encoded by this gene is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence. [provided by RefSeq, Jan 2012]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTAG1B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR