Anti-CTAG1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-4503R-A750
BOSSBS-4503R-A750EA 890 EUR
BOSSBS-4503R-A750
Anti-CTAG1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
The protein encoded by this gene is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence.

Type: Primary
Antigen: CTAG1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 81-180/180
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR