Anti-CTAG1B Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-4503R-CY5
BOSSBS-4503R-CY5EA 900 EUR
BOSSBS-4503R-CY5
Anti-CTAG1B Rabbit Polyclonal Antibody (Cy5®)
Antibodies
The protein encoded by this gene is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence. [provided by RefSeq, Jan 2012]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTAG1B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol CTAG1B
Environmentally Preferable
Antigen name Cancer/Testis Antigen 1B
Antigen synonyms LAGE2B|CT6.1|NY-ESO-1|CTAG1|CTAG|ESO1|LAGE-2
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human
Host Rabbit
Gene ID 1485
Isotype IgG
ImmunoChemistry Yes
Immunogen 100-150/180
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR