Anti-S. typhimurium Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Supplier: Bioss
BS-4801R-A647
BOSSBS-4801R-A647EA 893 EUR
BOSSBS-4801R-A647
Anti-S. typhimurium Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram-negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major antigens; the "H" or flagellar antigen (phase 1 & 2), the "O" or somatic antigen (part of the LPS moiety) and the "Vi" or capsular antigen (referred to as "K" in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram-negative bacteria. This LPS is composed of an "O" polysaccharide ("O" antigen) an "R" core and the endotoxic inner "Lipid A". Endotoxins evoke feverMore Product Information