Anti-WFDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-4838R-A647
BOSSBS-4838R-A647EA 1350 EUR
BOSSBS-4838R-A647
Anti-WFDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
HE4 is a broad range protease inhibitor. It is expressed in a number of normal tissues, including male reproductive system, regions of the respiratory tract and nasopharynx. It is highly expressed in a number of tumors cells lines, such ovarian, colon, breast, lung and renal cells lines.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WFDC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR