Anti-BAAT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Supplier: Bioss
BS-5040R-A647
BOSSBS-5040R-A647EA 893 EUR
BOSSBS-5040R-A647
Anti-BAAT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Involved in bile acid metabolism. In liver hepatocytes catalyzes the second step in the conjugation of C24 bile acids (choloneates) to glycine and taurine before excretion into bile canaliculi. The major components of bile are cholic acid and chenodeoxycholic acid. In a first step the bile acids are converted to an acyl-CoA thioester, either in peroxisomes (primary bile acids deriving from the cholesterol pathway), or cytoplasmic at the endoplasmic reticulum (secondary bile acids). May catalyze the conjugation of primary or secondary bile acids, or both. The conjugation increases theMore Product Information