Anti-FAAH2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-5119R-A350
BOSSBS-5119R-A350EA 893 EUR
BOSSBS-5119R-A350
Anti-FAAH2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
This gene encodes a fatty acid amide hydrolase that shares a conserved protein motif with the amidase signature family of enzymes. The encoded enzyme is able to catalyze the hydrolysis of a broad range of bioactive lipids, including those from the three main classes of fatty acid amides; N-acylethanolamines, fatty acid primary amides and N-acyl amino acids. This enzyme has a preference for monounsaturated acyl chains as a substrate.[provided by RefSeq, Sep 2009]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAAH2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 35More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR