Anti-BRK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
BS-5124R-HRP
BOSSBS-5124R-HRPEA 900 EUR
BOSSBS-5124R-HRP
Anti-BRK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
HSPC300 (hematopoietic stem cell protein 300) is also known as probable protein BRICK1 or C3orf10 (chromosome 3 open reading frame 10) and is a 75 amino acid protein that is expressed as two isoforms and localizes to both the cytoplasm and the cytoskeleton. HSPC300 is thought to regulate cytoskeletal organization and Actin polymerization. Free HSPC300 exists as homotrimers prior to its incorporation into the WAVE complex. The WAVE complex includes five proteins, one of which is HSPC300, that regulate the ARC (Arp2/3 complex) which is responsible for Actin nucleation and is Rac 1-dependent.More Product Information