Anti-AKT1/2/3 (Ser472/Ser473/Ser474) Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-5209R-A750
BOSSBS-5209R-A750EA 921 EUR
BOSSBS-5209R-A750
Anti-AKT1/2/3 (Ser472/Ser473/Ser474) Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
AKT3 is one of 3 closely related serine/threonine-protein kinases (AKT1, AKT2 and AKT3) called the AKT kinase, and which regulate many processes including metabolism, proliferation, cell survival, growth and angiogenesis. This is mediated through serine and/or threonine phosphorylation of a range of downstream substrates. Over 100 substrate candidates have been reported so far, but for most of them, no isoform specificity has been reported. AKT3 is the least studied AKT isoform. It plays an important role in brain development and is crucial for the viability of malignant glioma cells. AKT3More Product Information