Anti-PCDHGB5 Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-5827R-CY5.5
BOSSBS-5827R-CY5.5EA 900 EUR
BOSSBS-5827R-CY5.5
Anti-PCDHGB5 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
PCDHGB5 is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons sharedMore Product Information