Anti-CRABP2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-6038R-CY5.5
BOSSBS-6038R-CY5.5EA 900 EUR
BOSSBS-6038R-CY5.5
Anti-CRABP2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
A number of specific carrier proteins for members of the vitamin A family have been discovered. Cellular retinoic acid binding proteins (CRABP) are low molecular weight proteins whose precise function remains unknown. The inducibility of the CRABP2 gene suggests that this isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. It has been postulated that the CRABP2 gene is transcriptionally regulated by a newly synthesized regulatory protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRABP2
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR