Anti-RASSF8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-6089R-A750
BOSSBS-6089R-A750EA 893 EUR
BOSSBS-6089R-A750
Anti-RASSF8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
This gene encodes a member of the Ras-assocation domain family (RASSF) of tumor suppressor proteins. This gene is essential for maintaining adherens junction function in epithelial cells and has a role in epithelial cell migration. It is a lung tumor suppressor gene candidate. A chromosomal translocation t(12;22)(p11.2;q13.3) leading to the fusion of this gene and the FBLN1 gene is found in a complex type of synpolydactyly. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: RASSF8
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR