Anti-p73 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-6147R-A750
BOSSBS-6147R-A750EA 893 EUR
BOSSBS-6147R-A750
Anti-p73 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
This gene encodes a member of the p53 family of transcription factors involved in cellular responses to stress and development. It maps to a region on chromosome 1p36 that is frequently deleted in neuroblastoma and other tumors, and thought to contain multiple tumor suppressor genes. The demonstration that this gene is monoallelically expressed (likely from the maternal allele), supports the notion that it is a candidate gene for neuroblastoma. Many transcript variants resulting from alternative splicing and/or use of alternate promoters have been found for this gene, but the biologicalMore Product Information