Anti-LDHD Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-6272R-CY5.5
BOSSBS-6272R-CY5.5EA 900 EUR
BOSSBS-6272R-CY5.5
Anti-LDHD Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The protein encoded by this gene belongs to the D-isomer specific 2-hydroxyacid dehydrogenase family. The similar protein in yeast has both D-lactate and D-glycerate dehydrogenase activities. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq, Jul 2008]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LDHD
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR