Anti-CDK5 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-6411R-CY7
BOSSBS-6411R-CY7EA 902 EUR
BOSSBS-6411R-CY7
Anti-CDK5 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
CDK5 (Cyclin Dependent Kinase 5) is serine/threonine kinase involved in synaptic regulation and neuronal development; phosphorylates synaptic protein Pctaire1; regulates acetylcholine receptor expression. CDK5 is a member of the cyclindependent kinase family of serine/threonine kinases. It is present in numerous mammalian tissues including kidney, testes, and ovary. Its activity is detected almost exclusively in brain extracts. Neuronal and muscle cells contain the highest amount of this protein. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity, but Cdk5 is not activatedMore Product Information