Anti-CA8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-6516R-A680
BOSSBS-6516R-A680EA 893 EUR
BOSSBS-6516R-A680
Anti-CA8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.
Type: Primary