Anti-HRH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-6663R-A488
BOSSBS-6663R-A488EA 890 EUR
BOSSBS-6663R-A488
Anti-HRH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
In peripheral tissues, the H1 subclass of histamine receptors mediates the contraction of smooth muscles, increase in capillary permeability due to contraction of terminal venules, and catecholamine release from adrenal medulla, as well as mediating neurotransmission in the central nervous system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HRH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR