Anti-MAGEA10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-6816R-A555
BOSSBS-6816R-A555EA 893 EUR
BOSSBS-6816R-A555
Anti-MAGEA10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. Alternative splicing results in multiple transcript variants. Read-through transcription alsoMore Product Information