Anti-MAGEB2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
BS-6822R-HRP
BOSSBS-6822R-HRPEA 900 EUR
BOSSBS-6822R-HRP
Anti-MAGEB2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region. It is expressed in testis and placenta, and in a significant fraction of tumors of variousMore Product Information